MilliporeSigma
  • TG101209, a novel JAK2 inhibitor, has significant in vitro activity in multiple myeloma and displays preferential cytotoxicity for CD45+ myeloma cells.
TG101209, a novel JAK2 inhibitor, has significant in vitro activity in multiple myeloma and displays preferential cytotoxicity for CD45+ myeloma cells.
American journal of hematology (2010-07-24)
Vijay Ramakrishnan, Teresa Kimlinger, Jessica Haug, Michael Timm, Linda Wellik, Timothy Halling, Animesh Pardanani, Ayalew Tefferi, Sundararasan Vincent Rajkumar, Shaji Kumar
ABSTRACT
Interaction of myeloma cells with the bone marrow microenvironment is mediated in large part through different cytokines, especially VEGF and IL6. These cytokines, especially IL6, leads to upregulation of the JAK/STAT pathway in myeloma cell, contributing to increased proliferation, decreased apoptosis, and acquired drug resistance. Here, we examined the preclinical activity of a novel JAK2 inhibitor TG101209. TG101209 induced dose- and time-dependent cytotoxicity in a variety of multiple myeloma (MM) cell lines. The induction of cytotoxicity was associated with inhibition of cell cycle progression and induction of apoptosis in myeloma cell lines and patient-derived plasma cells. Evaluation of U266 cell lines and patient cells, which have a mix of CD45 positive and negative cells, demonstrated more profound cytotoxicity and antiproliferative activity of the drug on the CD45+ population relative to the CD45- cells. Exploring the mechanism of action of TG101209 indicated downregulation of pJak2, pStat3, and Bcl-xl levels with upregulation of pErk and pAkt levels indicating cross talk between signaling pathways. TG101209, when used in combination with the PI3K inhibitor LY294002, demonstrated synergistic cytotoxicity against myeloma cells. Our results provide the rationale for clinical evaluation of TG101209 alone or in combination with PI3K/Akt inhibitors in MM.
Sigma-Aldrich
Ribonuclease B from bovine pancreas, BioReagent, ≥50 Kunitz units/mg protein, ≥80% (SDS-PAGE)