MilliporeSigma
  • Liquid-phase combinatorial synthesis of 1,4-benzodiazepine-2,5-diones as the candidates of endothelin receptor antagonism.
Liquid-phase combinatorial synthesis of 1,4-benzodiazepine-2,5-diones as the candidates of endothelin receptor antagonism.
Journal of combinatorial chemistry (2004-01-13)
Ming-Fu Cheng, Jim-Min Fang
ABSTRACT
A library of 1,4-benzodiazepine-2,5-dione dicarboxylate derivatives containing aryl substituents at N(1)- and N(4)-positions to mimic the amino acid residues of Try-13, Phe-14, and Asp-18 in endothelin-1 is established by using the starting materials of alpha-amino esters, hydroxybenzaldehydes, nitrobenzoyl chlorides, and benzyl bromides in a polyethylene resin-bound liquid-phase synthesis. All of the six synthetic steps are conducted under mild conditions to give the desired products with reasonable yields and purity. The poly(ethylene glycol) support plays as a part of ester linkage that is released at the final step.