MilliporeSigma
  • Voltammetric determination of caffeine in beverages using a chemically modified electrode.
Voltammetric determination of caffeine in beverages using a chemically modified electrode.
The Analyst (1998-08-26)
J M Zen, Y S Ting, Y Shih
ABSTRACT
A Nafion-ruthenium oxide pyrochlore chemically modified electrode is used for the determination of caffeine in beverages by square-wave voltammetry. Compared to a bare glassy carbon electrode, the chemically modified electrode exhibits a marked enhancement of the current response. A linear calibration curve is obtained over the 5-200 microns range in 0.05 M HCIO4 solution with a detection limit (3 delta) of 2 micron. The results of 15 successive repetitive measurement-regeneration cycles showed a relative standard deviation of 2.7% for 10 micron caffeine. Thus, the electrode renewal gives a good reproducible surface. Quantitative analysis was performed by the standard addition method for caffeine content in tea, coffee, decaffeinated coffee, and cola beverages.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Pricing and availability is not currently available.