MilliporeSigma
  • Unique presence of a manganese catalase in a hyperthermophilic archaeon, Pyrobaculum calidifontis VA1.
Unique presence of a manganese catalase in a hyperthermophilic archaeon, Pyrobaculum calidifontis VA1.
Journal of bacteriology (2002-05-25)
Taku Amo, Haruyuki Atomi, Tadayuki Imanaka
ABSTRACT
We had previously isolated a facultatively anaerobic hyperthermophilic archaeon, Pyrobaculum calidifontis strain VA1. Here, we found that strain VA1, when grown under aerobic conditions, harbors high catalase activity. The catalase was purified 91-fold from crude extracts and displayed a specific activity of 23,500 U/mg at 70 degrees C. The enzyme exhibited a K(m) value of 170 mM toward H(2)O(2) and a k(cat) value of 2.9 x 10(4) s(-1).subunit(-1) at 25 degrees C. Gel filtration chromatography indicated that the enzyme was a homotetramer with a subunit molecular mass of 33,450 Da. The purified catalase did not display the Soret band, which is an absorption band particular to heme enzymes. In contrast to typical heme catalases, the catalase was not strongly inhibited by sodium azide. Furthermore, with plasma emission spectroscopy, we found that the catalase did not contain iron but instead contained manganese. Our biochemical results indicated that the purified catalase was not a heme catalase but a manganese (nonheme) catalase, the first example in archaea. Intracellular catalase activity decreased when cells were grown anaerobically, while under aerobic conditions, an increase in activity was observed with the removal of thiosulfate from the medium, or addition of manganese. Based on the N-terminal amino acid sequence of the purified protein, we cloned and sequenced the catalase gene (kat(Pc)). The deduced amino acid sequence showed similarity with that of the manganese catalase from a thermophilic bacterium, Thermus sp. YS 8-13. Interestingly, in the complete archaeal genome sequences, no open reading frame has been assigned as a manganese catalase gene. Moreover, a homology search with the sequence of kat(Pc) revealed that no orthologue genes were present on the archaeal genomes, including those from the "aerobic" (hyper)thermophilic archaea Aeropyrum pernix, Sulfolobus solfataricus, and Sulfolobus tokodaii. Therefore, Kat(Pc) can be considered a rare example of a manganese catalase from archaea.
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein