MilliporeSigma
  • Asymmetric hydrogenation of alpha-chloro aromatic ketones catalyzed by eta6-arene/TsDPEN-ruthenium(II) complexes.
Asymmetric hydrogenation of alpha-chloro aromatic ketones catalyzed by eta6-arene/TsDPEN-ruthenium(II) complexes.
Organic letters (2007-01-16)
Takeshi Ohkuma, Kunihiko Tsutsumi, Noriyuki Utsumi, Noriyoshi Arai, Ryoji Noyori, Kunihiko Murata
ABSTRACT
Asymmetric hydrogenation of various alpha-chloro aromatic ketones with Ru(OTf)(TsDPEN)(eta6-arene) (TsDPEN = N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine) produces the chiral chlorohydrins in up to 98% ee. This reaction can be conducted even on a 206-g scale. The hydrogenation of an alpha-chloro ketone with a phenol moiety has been utilized for the synthesis of (R)-norphenylephrine without protection-deprotection operations. [reaction: see text].