MilliporeSigma
  • Selective catalytic sp3 C-O bond cleavage with C-N bond formation in 3-alkoxy-1-propanols.
Selective catalytic sp3 C-O bond cleavage with C-N bond formation in 3-alkoxy-1-propanols.
Organic letters (2012-06-06)
Cheng Chen, Soon Hyeok Hong
ABSTRACT
The ruthenium catalyzed selective sp(3) C-O cleavage with amide formation was reported in reactions of 3-alkoxy-1-propanol derivatives and amines. The cleavage only occurs at the C3-O position even with 3-benzyloxy-1-propanol. Based on the experimental results, O-bound and C-bound Ru enolate complexes were proposed as key intermediates for the unique selective sp(3) C-O bond cleavage in 3-alkoxy-1-propanols.
Sigma-Aldrich
3-Benzyloxy-1-propanol, 97%