MilliporeSigma
  • Hydrogels for potential colon drug release by thiol-ene conjugate addition of a new inulin derivative.
Hydrogels for potential colon drug release by thiol-ene conjugate addition of a new inulin derivative.
Macromolecular bioscience (2008-06-26)
Giovanna Pitarresi, Giuseppe Tripodo, Rossella Calabrese, Emanuela Fabiola Craparo, Mariano Licciardi, Gaetano Giammona
ABSTRACT
Inulin was chosen as a starting polymer for biocompatible, pH-sensitive and biodegradable hydrogels. Three INUDVSA-TT hydrogels were obtained by crosslinking inulin derivatives with trimethylolpropane tris(3-mercaptopropionate) under varying conditions. The resulting hydrogels were cell compatible, as demonstrated by MTS and trypan blue exclusion assays acting on Caco-2 cells, and were biodegraded by inulinase and esterase, thus suggesting their use as colonic drug delivery systems. 2-Methoxyestradiol, an anti-cancer drug, was soaked in INUDVSA-TT hydrogels and its in vitro release and apoptotic effect on Caco-2 cells were evaluated.
Sigma-Aldrich
Trimethylolpropane tris(3-mercaptopropionate), ≥95.0%