MilliporeSigma
  • Antimalarials. 12. Preparation of carbon isosteres of selected 4-pyridinemethanols as suppressive antimalarials.
Antimalarials. 12. Preparation of carbon isosteres of selected 4-pyridinemethanols as suppressive antimalarials.
Journal of medicinal chemistry (1980-11-01)
A Markovac, M P LaMontagne
ABSTRACT
Four carbon isosteres related to the highly active 4-pyridylcarbinolamines were prepared and evaluated for suppressive antimalarial activity against Plasmodium berghei in mice. Three of the four examples possessed significant activity but were approximately one dose level less active than the corresponding pyridines.
Sigma-Aldrich
Methyl 3,5-dimethylbenzoate, 98%