MilliporeSigma
Enzymatic degradation of nitriles by a Candida guilliermondii UFMG-Y65.
Canadian journal of microbiology (2000-07-29)
J C Dias, R P Rezende, C A Rosa, M A Lachance, V R Linardi
ABSTRACT
Candida guilliermondii UFMG-Y65, isolated from a gold mine, was able to utilize different nitriles and the corresponding amides as sole source of nitrogen, at concentrations up to 2 M. Resting cells cultivated on YCB-acetonitrile medium showed nitrile hydrolyzing enzyme activities against acrylonitrile and benzonitrile. These enzymes were inducible and intracellular; the optimum pH was 7.0-8.0, and the optimum temperature 25 degrees C-30 degrees C. Liquid chromatographic analysis indicated that C. guilliermondii UFMG-Y65 metabolized 12 mM benzonitrile to 11 mM benzoic acid and 10 mM acrylonitrile to 7.9 mM acrylic acid. The results suggest that C. guilliermondii UFMG-Y65 may be useful for the bioproduction of amides and acids, and for the bioremediation of environments contaminated with nitriles.
Sigma-Aldrich
Cyclopentanecarbonitrile, 98%