MilliporeSigma
  • Molecular cloning and characterization of two novel retinoic acid-inducible orphan G-protein-coupled receptors (GPRC5B and GPRC5C).
Molecular cloning and characterization of two novel retinoic acid-inducible orphan G-protein-coupled receptors (GPRC5B and GPRC5C).
Genomics (2000-08-17)
M J Robbins, D Michalovich, J Hill, A R Calver, A D Medhurst, I Gloger, M Sims, D N Middlemiss, M N Pangalos
ABSTRACT
Using homology searching of public databases with a metabotropic glutamate receptor sequence from Caenorhabditis elegans, two novel protein sequences (named RAIG-2 (HGMW-approved symbol GPRC5B) and RAIG-3 (HGMW-approved symbol GPRC5C) were identified containing seven putative transmembrane domains characteristic of G-protein-coupled receptors (GPCRs). RAIG-2 and RAIG-3 encode open reading frames of 403 and 442 amino acid polypeptides, respectively, and show 58% similarity to the recently identified retinoic acid-inducible gene-1 (RAIG-1, HGMW-approved symbol RAI3). Analysis of the three protein sequences places them within the type 3 GPCR family, which includes metabotropic glutamate receptors, GABA(B) receptors, calcium-sensing receptors, and pheromone receptors. However, in contrast to other type 3 GPCRs, RAIG-1, RAIG-2, and RAIG-3 have only short N-terminal domains. RAIG-2 and RAIG-3 cDNA sequences were cloned into the mammalian expression vector pcDNA3 with c-myc or HA epitope tags inserted at their N-termini, respectively. Transient transfection experiments in HEK239T cells using these constructs demonstrated RAIG-2 and RAIG-3 expression at the cell surface. Distribution profiles of mRNA expression obtained by semiquantitative Taq-Man PCR analysis showed RAIG-2 to be predominantly expressed in human brain areas and RAIG-3 to be predominantly expressed in peripheral tissues. In addition, expression of RAIG-2 and RAIG-3 mRNA was increased following treatment with all-trans-retinoic acid in a manner similar to that previously described for RAIG-1. Finally, RAIG-2 was mapped to chromosome 16p12 (D16S405-D16S3045) and RAIG-3 to chromosome 17q25 (D17S1352-D17S785). These results suggest that RAIG-1, RAIG-2, and RAIG-3 represent a novel family of retinoic acid-inducible receptors, most closely related to the type 3 GPCR subfamily, and provide further evidence for a linkage between retinoic acid and G-protein-coupled receptor signal transduction pathways.