MilliporeSigma
  • The glucose-regulated proteins: stress induction and clinical applications.
The glucose-regulated proteins: stress induction and clinical applications.
Trends in biochemical sciences (2001-08-16)
A S Lee
ABSTRACT
A protective mechanism used by cells to adapt to stress of the endoplasmic reticulum (ER) is the induction of members of the glucose-regulated protein (Grp) family. The induction of mammalian Grp proteins in response to ER stress involves a complex network of regulators and novel mechanisms. The elucidation of Grp function and regulation opens up new therapeutic approaches to diseases associated with ER stress and cancer.