MilliporeSigma
  • Up-regulation of Flotillin-2 is associated with melanoma progression and modulates expression of the thrombin receptor protease activated receptor 1.
Up-regulation of Flotillin-2 is associated with melanoma progression and modulates expression of the thrombin receptor protease activated receptor 1.
Cancer research (2004-10-20)
Parul Hazarika, Marya F McCarty, Victor G Prieto, Saira George, Daniel Babu, Dimpy Koul, Menashe Bar-Eli, Madeleine Duvic
ABSTRACT
Flotillin 2 (flot-2) is a highly conserved protein isolated from caveolae/lipid raft domains that tether growth factor receptors linked to signal transduction pathways. Flot-2 protein and mRNA were increased in tumorigenic and metastatic melanoma cell lines in vitro, and the immunostaining intensity increased substantially across a tissue array of melanocytic lesions. Flot-2 transfection transformed SB2 melanoma cells from nontumorigenic, nonmetastatic to highly tumorigenic and metastatic in a nude mouse xenograft model. SB2 cells stably transfected with the flot-2 cDNA (SB2-flot)-2 cells proliferated faster in the absence of serum, and their migration through Matrigel was additionally enhanced by thrombin. When SB2-flot-2 cells were compared with SB2-vector-control cells on a cancer gene pathway array, SB2-flot-2 cells had increased expression of protease activated receptor 1 (PAR-1) mRNA, a transmembrane, G-protein-coupled receptor involved in melanoma progression. PAR-1 and flot-2 were coimmunoprecipitated from SB2-flot-2 cells. Up-regulation of PAR-1 was additionally confirmed in SB2-flot-2 cells and melanoma cell lines. SB2-flot-2 cells transfected with flot-2-specific small-interfering RNAs made substantially less flot-2 and PAR-1 mRNA. In conclusion, flot-2 overexpression is associated with melanoma progression, with increased PAR-1 expression, and with transformation of SB2 melanoma cells to a highly metastatic line. Flot-2 binds to PAR-1, a known upstream mediator of major signal transduction pathways implicated in cell growth and metastasis, and may thereby influence tumor progression.