MilliporeSigma
Regioselective synthesis of beta-N1- and beta-N3-alloxazine nucleosides
Organic letters (2000-05-18)
Wang, Rizzo
ABSTRACT
[structures: see text] A regio- and stereoselective glycosylation of ribose tetraester with persilylated alloxazine to give either beta-N1 or beta-N3 nucleosides is described. The N3 product is potentially of interest as a fluorescent nucleoside and is predicted to have the hydrogen-bonding characteristics of thymidine.