MilliporeSigma
  • The human homolog of the mouse common viral integration region, FLI1, maps to 11q23-q24.
The human homolog of the mouse common viral integration region, FLI1, maps to 11q23-q24.
Genomics (1991-09-01)
V Baud, M Lipinski, E Rassart, L Poliquin, D Bergeron
ABSTRACT
FLI1 is a common mouse viral integration region in virus-induced leukemias and lymphomas. Using an evolutionarily conserved mouse probe and Southern hybridization to (rodent x human) somatic cell hybrid DNAs, the human homolog of FLI1 has been shown to lie on a fragment of chromosome 11 flanked on the centromeric side by the acute lymphoblastic leukemia-associated t(4;11)(q21;q23) translocation breakpoint and on the telomeric side by the Ewing- and neuroepithelioma-associated t(11;22) (q24;q12) breakpoint.