MilliporeSigma
  • Preparation of Weinreb amides using 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride (DMT-MM).
Preparation of Weinreb amides using 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride (DMT-MM).
Chemical & pharmaceutical bulletin (2004-04-02)
Kazuhito Hioki, Hiroko Kobayashi, Rumi Ohkihara, Shohei Tani, Munetaka Kunishima
ABSTRACT
Weinreb amides were successfully prepared from the corresponding carboxylic acids using 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride (DMT-MM) in the solvents, methanol, isopropyl alcohol, and acetonitrile, which can solubilize DMT-MM. A variety of carboxylic acids were converted to the corresponding Weinreb amides in excellent yields by simply mixing with DMT-MM and N,O-dimethylhydroxylamine hydrochloride.
Sigma-Aldrich
N-Methoxy-N-methylbenzamide, 98%
Sigma-Aldrich
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride, 95% (HPLC)