MilliporeSigma
  • Effect on cardiovascular risk of high density lipoprotein targeted drug treatments niacin, fibrates, and CETP inhibitors: meta-analysis of randomised controlled trials including 117,411 patients.
Effect on cardiovascular risk of high density lipoprotein targeted drug treatments niacin, fibrates, and CETP inhibitors: meta-analysis of randomised controlled trials including 117,411 patients.
BMJ (Clinical research ed.) (2014-07-20)
Daniel Keene, Clare Price, Matthew J Shun-Shin, Darrel P Francis
ABSTRACT
To investigate the effects on cardiovascular outcomes of drug interventions that increase high density lipoprotein levels. Meta-analysis. Therapeutic benefit of niacin, fibrates, and cholesteryl ester transfer protein (CETP) inhibitors on cardiovascular events (all cause mortality, coronary heart disease mortality, non-fatal myocardial infarction, and stroke). 117,411 patients were randomised in a total of 39 trials. All interventions increased the levels of high density lipoprotein cholesterol. No significant effect was seen on all cause mortality for niacin (odds ratio 1.03, 95% confidence interval 0.92 to 1.15, P=0.59), fibrates (0.98, 0.89 to 1.08, P=0.66), or CETP inhibitors (1.16, 0.93 to 1.44, P=0.19); on coronary heart disease mortality for niacin (0.93, 0.76 to 1.12, P=0.44), fibrates (0.92, 0.81 to 1.04, P=0.19), or CETP inhibitors (1.00, 0.80 to 1.24, P=0.99); or on stroke outcomes for niacin (0.96, 0.75 to 1.22, P=0.72), fibrates (1.01, 0.90 to 1.13, P=0.84), or CETP inhibitors (1.14, 0.90 to 1.45, P=0.29). In studies with patients not receiving statins (before the statin era), niacin was associated with a significant reduction in non-fatal myocardial infarction (0.69, 0.56 to 0.85, P=0.0004). However, in studies where statins were already being taken, niacin showed no significant effect (0.96, 0.85 to 1.09, P=0.52). A significant difference was seen between these subgroups (P=0.007). A similar trend relating to non-fatal myocardial infarction was seen with fibrates: without statin treatment (0.78, 0.71 to 0.86, P<0.001) and with all or some patients taking statins (0.83, 0.69 to 1.01, P=0.07); P=0.58 for difference. Neither niacin, fibrates, nor CETP inhibitors, three highly effective agents for increasing high density lipoprotein levels, reduced all cause mortality, coronary heart disease mortality, myocardial infarction, or stroke in patients treated with statins. Although observational studies might suggest a simplistic hypothesis for high density lipoprotein cholesterol, that increasing the levels pharmacologically would generally reduce cardiovascular events, in the current era of widespread use of statins in dyslipidaemia, substantial trials of these three agents do not support this concept.
Sigma-Aldrich
Torcetrapib, ≥98% (HPLC)
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid, SAJ special grade, ≥99.5%
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinic acid sodium salt, 98%
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Nicotinic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Niacin, United States Pharmacopeia (USP) Reference Standard