MilliporeSigma
  • Chemical modification of temoporfin--a second generation photosensitizer activated using upconverting nanoparticles for singlet oxygen generation.
Chemical modification of temoporfin--a second generation photosensitizer activated using upconverting nanoparticles for singlet oxygen generation.
Chemical communications (Cambridge, England) (2014-09-02)
Q Yu, E M Rodriguez, R Naccache, P Forgione, G Lamoureux, F Sanz-Rodriguez, D Scheglmann, J A Capobianco
ABSTRACT
LiYF4:Tm(3+)/Yb(3+) upconverting nanoparticles (UCNPs) were functionalized with the second generation photosensitizer 5,10,15,20-tetra(m-hydroxyphenyl)chlorin (m-THPC, Temoporfin, Foscan®). m-THPC was modified using 4-(bromomethyl)benzoic acid, which induced a bathochromic shift of the m-THPC blue absorption peak. The nanoconstruct causes up to 70% cell death under 980 nm irradiation.
Sigma-Aldrich
4-(Bromomethyl)benzoic acid, 97%