MilliporeSigma
  • Synthesis of oligosaccharides corresponding to Vibrio cholerae O139 polysaccharide structures containing dideoxy sugars and a cyclic phosphate.
Synthesis of oligosaccharides corresponding to Vibrio cholerae O139 polysaccharide structures containing dideoxy sugars and a cyclic phosphate.
Organic & biomolecular chemistry (2006-03-25)
Dominika Turek, Andreas Sundgren, Martina Lahmann, Stefan Oscarson
ABSTRACT
A spacer-equipped tetrasaccharide, p-aminocyclohexylethyl alpha-l-Colp-(1-->2)-beta-d-Galp-(1-->3)-[alpha-l-Colp-(1-->4)]-beta-D-GlcpNAc, containing a 4,6-cyclic phosphate in the galactose residue, has been synthesised. The structure corresponds to a part of the repeating unit of the capsular (and lipo-) polysaccharide of the endemic bacteria Vibrio cholerae type O139 synonym Bengal. The synthetic strategy allows continuous syntheses of the complete O139 hexasaccharide repeating unit as well as of the structurally related repeating unit of serotype O22. Starting from ethyl 2-azido-4,6-O-benzylidene-2-deoxy-1-thio-beta-D-glucopyranoside, a thioglycoside tetrasaccharide donor block was constructed through two orthogonal glycosylations with glycosyl bromide donors. First, a properly protected galactose moiety was introduced using silver triflate as promoter and subsequently the two colitose residues, carrying electron-withdrawing protecting groups for stability reasons, under halide-assisted conditions. The tetrasaccharide block was then linked to the spacer in a NIS-TMSOTf-promoted coupling. Transformation of the azido group into an acetamido group using H2S followed by removal of temporary protecting acetyl groups gave a 4',6'-diol, which was next phosphorylated with methyl dichlorophosphate and deprotected to yield the 4,6-cyclic phosphate tetrasaccharide target structure.
Sigma-Aldrich
Methyl dichlorophosphate, 85%