MilliporeSigma
  • Cloning and sequence analysis of two laccase complementary DNAs from the ligninolytic basidiomycete Trametes versicolor.
Cloning and sequence analysis of two laccase complementary DNAs from the ligninolytic basidiomycete Trametes versicolor.
Gene (1997-10-10)
E Ong, W B Pollock, M Smith
ABSTRACT
Laccases are oxidoreductase enzymes involved in the oxidation of various phenolic compounds. They may play a role in the biodegradation of lignin and in the dechlorination of chlorophenols. The cDNAs encoding laccase LccI and a putative laccase LccIV and the gene for LccI from the white-rot basidiomycete Trametes versicolor were cloned, sequenced and characterized. The genomic DNA of lccI consists of 2128 bp, with the coding region interrupted by 10 introns; the cDNA consists of a 1560 bp open reading frame (ORF). The cDNA of the putative lccIV gene consists of a 1581 bp ORF, with a 794 bp 5' untranslated region. The size of the major transcript for both lccI and lccIV is approximately 2.3 kb. Transcription of lccIV was induced by 2,5-dimethylaniline, whereas the opposite effect was observed for lccI. Laccases I and IV contain highly conserved histidinyl and cysteinyl residues, believed to be involved in binding copper, and share extensive sequence similarity with other laccases produced by both ligninolytic and non-ligninolytic fungi.
Sigma-Aldrich
2,5-Dimethylaniline, 99%