MilliporeSigma
  • Modulation of ara-C induced apoptosis in leukemia by the PKC activator bryostatin 1.
Modulation of ara-C induced apoptosis in leukemia by the PKC activator bryostatin 1.
Frontiers in bioscience : a journal and virtual library (1997-06-01)
S Grant
ABSTRACT
Modulation of ara-C-induced apoptosis in human leukemia cells by the macrocyclic lactone PKC activator bryostatin 1 occurs at multiple levels, and involves a variety of oncogenes and signalling pathways. Under some circumstances, bryostatin 1 may lead to enhanced conversion of ara-C to its lethal metabolite, ara-CTP. However, bryostatin 1 is able to potentiate ara-C-mediated cytotoxicity in the absence of metabolic perturbations, presumably by modulating the cell death pathway itself. For example, chronic exposure of cells to bryostatin 1 leads to PKC down-regulation, which may alter the balance between survival (e.g., ERK) versus stress (e.g., SAPK/JNK)-related pathways. The ability of bryostatin 1 to enhance ara-C-mediated apoptosis is inversely related to its capacity to induce leukemic cell maturation and may involve the failure to down-regulate expression of the cell cycle progression-related proto-oncogene, c-myc. Finally, recent evidence suggests that bryostatin 1 may act, through modification of Bcl-2 phosphorylation status, at a distal site in the cell death pathway. These studies could provide a paradigm important for understanding the mechanism(s) by which agents acting through signal transduction pathways modulate cytotoxic drug-induced cell death
Sigma-Aldrich
Bryostatin 1, ≥99%, solid