MilliporeSigma
  • Thymoquinone: fifty years of success in the battle against cancer models.
Thymoquinone: fifty years of success in the battle against cancer models.
Drug discovery today (2013-09-05)
Regine Schneider-Stock, Isabelle H Fakhoury, Angela M Zaki, Chirine O El-Baba, Hala U Gali-Muhtasib
ABSTRACT
Thymoquinone (TQ), the main active constituent of black seed essential oil, exhibits promising effects against inflammatory diseases and cancer. TQ, modulates signaling pathways that are key to cancer progression, and enhances the anticancer potential of clinical drugs while reducing their toxic side effects. Considering that TQ was isolated 50 years ago, this review focuses on TQ's chemical and pharmacological properties and the latest advances in TQ analog design and nanoformulation. We discuss our current state of knowledge of TQ's adjuvant potential and in vivo antitumor activity and highlight its ability to modulate the hallmarks of cancer.
Sigma-Aldrich
Thymoquinone, ≥98%
Pricing and availability is not currently available.