MilliporeSigma
  • Mechanistic studies on the reactions of platinum(II) complexes with nitrogen- and sulfur-donor biomolecules.
Mechanistic studies on the reactions of platinum(II) complexes with nitrogen- and sulfur-donor biomolecules.
Dalton transactions (Cambridge, England : 2003) (2012-08-15)
Živadin D Bugarčić, Jovana Bogojeski, Biljana Petrović, Stephanie Hochreuther, Rudi van Eldik
ABSTRACT
A brief overview of mechanistic studies on the reactions of different Pt(II) complexes with nitrogen- and sulfur-donor biomolecules is presented. The first part describes the results obtained for substitution reactions of mono-functional Pt(II) complexes with different biomolecules, under various experimental conditions (temperature, pH and ionic strength). In addition, an overview of the results obtained for the substitution reactions of bi-functional Pt(II) complexes, analogous to cisplatin, with biomolecules is given. The last part of this report deals with different polynuclear Pt(II) complexes and their substitution behaviour with different biomolecules. The purpose of this perspective is to improve the understanding of the mechanism of action of Pt(II) complexes as potential anti-tumour drugs in the human body.
Sigma-Aldrich
Platinum black, black, powder, ≤20 μm, ≥99.95% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum black, low bulk density, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, gauze, 100 mesh, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum black, fuel cell grade, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.076 mm, ≥99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 6mm disks, thickness 0.025mm, as rolled, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 0.2m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 50x50mm, thickness 0.1mm, as rolled, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 10mm disks, thickness 0.0125mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, nanopowder, 200 nm particle size (SEM), 99.9% (metals basis)
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 0.2m coil, thickness 0.045mm, coil width .75mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 10mm disks, thickness 0.0075mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 0.05m coil, thickness 0.025mm, coil width .4mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 6mm disks, thickness 0.005mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.998%
SKU
Pack Size
Availability
Price
Quantity
Platinum, wire reel, 0.1m, diameter 0.15mm, annealed, may be used as one component of a thermocouple, 99.99%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 8mm disks, thickness 0.005mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, wire reel, 0.1m, diameter 0.006mm, wollaston, 99.9%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 0.1m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 4mm disks, thickness 0.025mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 15mm disks, thickness 0.025mm, as rolled, 99.95%