MilliporeSigma
Methanol and ethylene glycol intoxication.
Critical care clinics (2012-09-25)
James A Kruse
ABSTRACT
Accidental or intentional ingestion of substances containing methanol and ethylene glycol can result in death, and some survivors are left with blindness, renal dysfunction, and chronic brain injury. However, even in large ingestions, a favorable outcome is possible if the patient arrives at the hospital early enough and the poisoning is identified and appropriately treated in a timely manner. This review covers the common circumstances of exposure, the involved toxic mechanisms, and the clinical manifestations, laboratory findings, and treatment of methanol and ethylene glycol intoxication.
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Ethylene glycol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Pricing and availability is not currently available.
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Pricing and availability is not currently available.
Supelco
Methanol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, SAJ special grade
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, NMR reference standard
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, Absolute - Acetone free
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Pricing and availability is not currently available.