MilliporeSigma
  • [Bismuth preparations in the treatment of patients with gastric and duodenal ulcer disease].
[Bismuth preparations in the treatment of patients with gastric and duodenal ulcer disease].
Klinicheskaia meditsina (2008-12-04)
I V Maev, A A Samsonov, N N Golubev
ABSTRACT
Studies of the role of H. pylori (Hp) in etiopathogenesis of a number of common diseases of the upper gastrointestinal tract changed traditional approaches to their management with emphasis on antibiotic therapy. Positive effect of Hp eradication was confirmed in numerous randomized clinical studies. Bismuth-containing drugs have been used for a long time to treat gastrointestinal disorders. To-day, bismuth tri-potassium di-citrate (de---nol) is considered to be an optimal tool for different schemes of eradication therapy (triple and quadruple therapy of the first and second lines) and recommended to improve its efficiency by overcoming resistance of Hp to antibiotics. The overall efficacy of anti-helicobacterial therapy using bismuth tri-potassium di-citrate amounts to 81-98%. This compound has cytoprotective effect on gastric and duodenal mucosa, besides antibacterial action. All these properties characterize de-nol as a single universal preparation for the treatment of erosive ulcers in the gastrointestinal tract.
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Bismuth, foil, 25x25mm, thickness 0.5mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 25x25mm, thickness 1.0mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 25x25mm, thickness 2.0mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 150x150mm, thickness 0.025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 50x50mm, thickness 0.25mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 150x150mm, thickness 0.02mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 50x50mm, thickness 0.5mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, beads, 1-5 mm, 99.999% trace metals basis
Pricing and availability is not currently available.
Bismuth, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 150x150mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 50x50mm, thickness 1.0mm, 100%
Pricing and availability is not currently available.