MilliporeSigma
  • Spotlight on HDL-raising therapies: insights from the torcetrapib trials.
Spotlight on HDL-raising therapies: insights from the torcetrapib trials.
Nature clinical practice. Cardiovascular medicine (2008-04-24)
Anatol Kontush, Maryse Guérin, M John Chapman
ABSTRACT
Subnormal levels of HDL cholesterol constitute a major cardiovascular risk factor. Inhibitors of cholesteryl ester transfer protein (CETP) are presently the most potent HDL-raising agents. Torcetrapib was the first CETP inhibitor to enter a large-scale, prospective, placebo-controlled interventional trial, which was prematurely terminated in December 2006 because of excess cardiovascular and noncardiovascular mortality in the active treatment group. Therapy with torcetrapib was associated with considerable increases in aldosterone level and blood pressure and changes in serum electrolytes indicative of mineralocorticoid excess. These findings indicate that torcetrapib has off-target toxic effects unrelated to HDL raising that involve the activation of mineralocorticoid receptors by aldosterone and result in the induction of hypertension. In contrast with torcetrapib, other CETP inhibitors such as JTT-705 and MK-825 do not increase blood pressure in humans, an observation which discounts a class effect. The available data do not, however, exclude potential adverse effects of CETP inhibition such as the generation of HDL particles that have deficient biological activities and a deleterious impact on reverse cholesterol transport and steroid metabolism. Normalization of both defective HDL function and diminished HDL levels should, therefore, be the focus of pharmacological HDL raising in future studies.
Sigma-Aldrich
Torcetrapib, ≥98% (HPLC)