MilliporeSigma
  • Etodolac. A reappraisal of its pharmacology and therapeutic use in rheumatic diseases and pain states.
Etodolac. A reappraisal of its pharmacology and therapeutic use in rheumatic diseases and pain states.
Drugs (1991-08-01)
J A Balfour, M M Buckley
ABSTRACT
Etodolac is a nonsteroidal anti-inflammatory drug (NSAID) effective in the treatment of rheumatoid arthritis, osteoarthritis and ankylosing spondylitis, and in the alleviation of postoperative pain. Etodolac also provides relief of other types of pain, including that arising from gouty conditions and traumatic injury. In all indications, etodolac appears to be at least as effective as other NSAIDs. The incidence of clinical adverse effects other than abdominal pain and dyspepsia is similar to that observed with placebo, and etodolac has been associated with a low rate of gastrointestinal ulceration and other serious events. Data from preliminary animal studies have suggested that etodolac may provide more selective inhibition of prostaglandin synthesis at sites of inflammation than some other currently available NSAIDs. Thus, available evidence indicates that etodolac, with its low incidence of gastrointestinal events, is an effective and well tolerated alternative to other NSAIDs in the treatment of arthritic diseases and pain of various aetiologies and should be considered a first-line therapy.
Sigma-Aldrich
Etodolac
Etodolac, European Pharmacopoeia (EP) Reference Standard