MilliporeSigma
Bryostatin-1: a novel PKC inhibitor in clinical development.
Cancer investigation (2004-01-23)
Jeremy Kortmansky, Gary K Schwartz
ABSTRACT
Modulation of PKC represents a novel approach to cancer therapy. Bryostatin-1 is a macrocyclic lactone derived from a marine invertebrate that binds to the regulatory domain of protein kinase C. Short-term exposure to bryostatin-1 promotes activation of PKC, whereas prolonged exposure promotes significant downregulation of PKC. In numerous hematological and solid tumor cell lines, bryostatin-1 inhibits proliferation, induces differentiation, and promotes apoptosis. Furthermore, preclinical studies indicate that bryostatin-1 potently enhances the effect of chemotherapy. In many cases, this effect is sequence specific. Bryostatin-1 is currently in phase I and phase II clinical trials. The major toxicities are myalgias, nausea, and vomiting. Although there is minimal single-agent activity, combinations with standard chemotherapy are providing very encouraging results and indicate a new direction in cancer therapy.
Sigma-Aldrich
Bryostatin 1, ≥99%, solid