MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Ciclopirox for the treatment of superficial fungal infections: a review.
Ciclopirox for the treatment of superficial fungal infections: a review.
International journal of dermatology (2003-08-05)
Aditya K Gupta, Alayne R Skinner
ABSTRACT
Ciclopirox is a broad-spectrum antifungal agent that also exhibits anti-inflammatory and antibacterial activity. The lotion and cream formulations of ciclopirox are effective in many types of infection, including tinea corporis/cruris, tinea pedis, cutaneous candidiasis, pityriasis (tinea) versicolor, and seborrheic dermatitis. The new ciclopirox gel 0.77% formulation is also indicated for the treatment of seborrheic dermatitis of the scalp, interdigital tinea pedis and tinea corporis.
Product Description
Ciclopirox, European Pharmacopoeia (EP) Reference Standard