MilliporeSigma
  • CYP 17 and CYP 19 inhibitors. Evaluation of fluorine effects on the inhibiting activity of regioselectively fluorinated 1-(Naphthalen-2-ylmethyl)imidazoles.
CYP 17 and CYP 19 inhibitors. Evaluation of fluorine effects on the inhibiting activity of regioselectively fluorinated 1-(Naphthalen-2-ylmethyl)imidazoles.
Journal of enzyme inhibition and medicinal chemistry (2004-09-29)
Rolf W Hartmann, Anja Palusczak, Fabrice Lacan, Giacomo Ricci, Renzo Ruzziconi
ABSTRACT
Regioselectively fluorinated 1-(naphth-2-ylmethyl)imidazoles 1a-h have been synthesized starting from the corresponding (naphth-2-yl)methanols (2). 2a-d have been obtained by LiAlH4-promoted reduction of fluorinated 1-methyl-2-naphthaldehydes. The latter were easily prepared in fairly good overall yields by ceric ammonium nitrate (CAN)-promoted oxidative addition of the suitable 3-(fluoroaryl)-1-trimethylsilyloxy-1-butenes to ethyl vinyl ether in methanol followed by cyclization of the resulting acetals in strongly acidic medium in the presence of DDQ. 2e-h were prepared by LiAlH4-promoted reduction of the corresponding fluorinated methyl 2-naphthoates. The latter were more profitably obtained by reacting the suitable benzyl bromide with the sodium salt of dimethyl 2-(2,2-dimethoxyethyl)malonate in DMF followed by demethoxycarbonylation and acid catalysed cyclization of the resulting acetals. Compared with the nonfluorinated parent compounds 1i-1, fluorinated 1-(naphth-2-yl)methylimidazoles 1a-h turned out to be potent inhibitors of CYP17 and CYP19 enzymes. The most active inhibitor of CYP17 is 1c, whereas CYP19 is strongly inhibited by 1b, 1e, and 1g. Interestingly, 1g is a potent dual inhibitor also being very active towards CYP19.