MilliporeSigma
  • Synthesis of 3-substituted benzamides and 5-substituted isoquinolin-1(2H)-ones and preliminary evaluation as inhibitors of poly(ADP-ribose)polymerase (PARP).
Synthesis of 3-substituted benzamides and 5-substituted isoquinolin-1(2H)-ones and preliminary evaluation as inhibitors of poly(ADP-ribose)polymerase (PARP).
Bioorganic & medicinal chemistry (1998-07-29)
C Y Watson, W J Whish, M D Threadgill
ABSTRACT
Inhibitors of poly(ADP-ribose)polymerase (PARP) inhibit repair of damaged DNA and thus potentiate radiotherapy and chemotherapy of cancer. 3-Substituted benzamides and 5-substituted isoquinolin-1-ones have been synthesised and evaluated for inhibition of PARP. Reduction of 3-(bromoacetyl)benzamide, followed by treatment with base, gave RS-3-oxiranylbenzamide. Reduction of 3-(hydroxyacetyl)benzonitrile with bakers' yeast gave the R-diol which was converted to R-3-(1,2-dihydroxyethyl)benzamide. Similar reduction of 3-(acetoxyacetyl)benzonitrile led towards the S-diol which was converted to its cyclic acetonide. E-2-(2,6-Dicyanophenyl)-N,N-dimethylethenamine was formed by condensation of 2,6-dicyanotoluene with dimethylformamide dimethyl acetal (DMFDMA); cyclisation under acidic conditions afforded 5-cyanoisoquinolin-1-one. Heck coupling of 5-iodoisoquinolin-1-one with propenoic acid formed E-3-(1-oxoisoquinolin-5-yl)propenoic acid. 3-Oxiranylbenzamide, 5-bromoisoquinolin-1-one and 5-iodoisoquinolin-1-one were among the most potent inhibitors of PARP activity in a preliminary screen in vitro.