MilliporeSigma
  • Haloperoxidases: Enzymatic Synthesis of alpha,beta-Halohydrins from Gaseous Alkenes.
Haloperoxidases: Enzymatic Synthesis of alpha,beta-Halohydrins from Gaseous Alkenes.
Applied and environmental microbiology (1983-02-01)
J Geigert, S L Neidleman, D J Dalietos, S K Dewitt
ABSTRACT
The enzymatic synthesis of alpha,beta-halohydrins from gaseous alkenes is described. The enzymatic reaction required an alkene, a halide ion, dilute hydrogen peroxide, and a haloperoxidase enzyme. A wide range of gaseous alkenes were suitable for this reaction, including those containing isolated, conjugated, and cumulative carbon-carbon double bonds. Chlorohydrins, bromohydrins, and iodohydrins could be formed. The combining of this enzymatic synthesis with a previously described enzymatic synthesis of epoxides from alpha,beta-halohydrins provides an alternate pathway, other than the well-known enzymatic direct epoxidation pathway, from alkene to an epoxide.
Sigma-Aldrich
1-Chloro-2-propanol, 70%