MilliporeSigma
  • Selective proliferation of intestinal Barnesiella under fucosyllactose supplementation in mice.
Selective proliferation of intestinal Barnesiella under fucosyllactose supplementation in mice.
The British journal of nutrition (2014-01-15)
Gisela A Weiss, Christophe Chassard, Thierry Hennet
ABSTRACT
The oligosaccharides 2-fucosyllactose and 3-fucosyllactose are major constituents of human breast milk but are not found in mouse milk. Milk oligosaccharides have a prebiotic action, thus affecting the colonisation of the infant intestine by microbiota. To determine the specific effect of fucosyllactose exposure on intestinal microbiota in mice, in the present study, we orally supplemented newborn mice with pure 2-fucosyllactose and 3-fucosyllactose. Exposure to 2-fucosyllactose and 3-fucosyllactose increased the levels of bacteria of the Porphyromonadaceae family in the intestinal gut, more precisely members of the genus Barnesiella as analysed by 16S pyrosequencing. The ability of Barnesiella to utilise fucosyllactose as energy source was confirmed in bacterial cultures. Whereas B. intestinihominis and B. viscericola did not grow on fucose alone, they proliferated in the presence of 2-fucosyllactose and 3-fucosyllactose following the secretion of linkage-specific fucosidase enzymes that liberated lactose. The change in the composition of intestinal microbiota mediated by fucosyllactose supplementation affected the susceptibility of mice to dextran sulphate sodium-induced colitis, as indicated by increased resistance of mice subjected to 2-fucosyllactose supplementation for 6 weeks. The present study underlines the ability of specific milk oligosaccharides to change the composition of intestinal microbiota and thereby to shape an intestinal milieu resilient to inflammatory diseases.
Product Description
USP
Anhydrous lactose, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Pricing and availability is not currently available.
Lactose (anhydrous), European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Lactose, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.