- Chemically derived graphene oxide: towards large-area thin-film electronics and optoelectronics.
Chemically derived graphene oxide: towards large-area thin-film electronics and optoelectronics.
Advanced materials (Deerfield Beach, Fla.) (2010-05-01)
Goki Eda, Manish Chhowalla
PMID20432408
ABSTRACT
Chemically derived graphene oxide (GO) possesses a unique set of properties arising from oxygen functional groups that are introduced during chemical exfoliation of graphite. Large-area thin-film deposition of GO, enabled by its solubility in a variety of solvents, offers a route towards GO-based thin-film electronics and optoelectronics. The electrical and optical properties of GO are strongly dependent on its chemical and atomic structure and are tunable over a wide range via chemical engineering. In this Review, the fundamental structure and properties of GO-based thin films are discussed in relation to their potential applications in electronics and optoelectronics.