MilliporeSigma
  • Dose-response associations of silica with nonmalignant respiratory disease and lung cancer mortality in the diatomaceous earth industry.
Dose-response associations of silica with nonmalignant respiratory disease and lung cancer mortality in the diatomaceous earth industry.
American journal of epidemiology (1997-04-15)
H Checkoway, N J Heyer, N S Seixas, E A Welp, P A Demers, J M Hughes, H Weill
ABSTRACT
The potential carcinogenicity of crystalline silica to humans remains a controversial issue. The authors conducted an historical cohort mortality study of 2,342 male workers exposed to crystalline silica, predominantly cristobalite, in a diatomaceous earth mining and processing facility in California. During the years 1942-1994, mortality excesses were detected for nonmalignant respiratory diseases (NMRD) (standardized mortality ratio = 2.01, 95% confidence interval (CI) 1.56-2.55) and lung cancer (standardized mortality ratio = 1.29, 95% CI 1.01-1.61). NMRD mortality rose sharply with cumulative exposure to respirable crystalline silica; allowing for a 15-year latency, the rate ratio for the highest exposure stratum (> or = 5.0 mg/m3-years) was 5.35 (95% CI 2.23-12.8). The rate ratio for lung cancer reached 2.15 (95% CI 1.08-4.28) in the highest exposure category. These associations were unlikely to have been confounded by smoking or asbestos exposure. The findings indicate a strong dose-response relation for crystalline silica and NMRD mortality. The lung cancer results, although less convincing, add further support to an etiologic role for crystalline silica.
Product Description
Supelco
Celite® 545 AW, reagent grade