MilliporeSigma
  • Borrowing hydrogen: iridium-catalysed reactions for the formation of C-C bonds from alcohols.
Borrowing hydrogen: iridium-catalysed reactions for the formation of C-C bonds from alcohols.
Organic & biomolecular chemistry (2005-12-17)
Phillip J Black, Gerta Cami-Kobeci, Michael G Edwards, Paul A Slatford, Michael K Whittlesey, Jonathan M J Williams
ABSTRACT
Alcohols have been employed as substrates for C-C bond-forming reactions which involve initial activation by the temporary removal of hydrogen to form an aldehyde. The intermediate aldehyde is converted into an alkene via a Horner-Wadsworth-Emmons reaction, nitroaldol and aldol reactions. The 'borrowed hydrogen' is then returned to the alkene to form a C-C bond.