MilliporeSigma
3-Bromo-N'-(2-hydroxy-benzyl-idene)benzohydrazide.
Acta crystallographica. Section E, Structure reports online (2008-01-01)
He-Bing Li
ABSTRACT
The title mol-ecule, C(14)H(11)BrN(2)O(2), displays a trans configuration about the C=N and C-N bonds. The dihedral angle between the two benzene rings is 18.5 (3)°. An intra-molecular O-H⋯N hydrogen bond is observed. In the crystal structure, the mol-ecules are linked into a chain along the c axis by N-H⋯O and C-H⋯O hydrogen bonds.