MilliporeSigma
Iridoids from Teucrium yemense.
Archives of pharmacal research (1998-12-30)
E Abdel-Sattar
ABSTRACT
The aerial parts of Teucrium yemense yielded two iridoid glycosides. Their structures were elucidated by spectral means as teucardosid and 8-O-acetylharpagid.
Sigma-Aldrich
8-Acetylharpagide, ≥85% (LC/MS-ELSD)