MilliporeSigma
  • Evaluation of ability of ferulic acid to control growth and fumonisin production of Fusarium verticillioides and Fusarium proliferatum on maize based media.
Evaluation of ability of ferulic acid to control growth and fumonisin production of Fusarium verticillioides and Fusarium proliferatum on maize based media.
International journal of food microbiology (2013-10-22)
Laura Ferrochio, Eugenia Cendoya, María Cecilia Farnochi, Walter Massad, María Laura Ramirez
ABSTRACT
The aim of this study was to evaluate the efficacy of ferulic acid (1, 10, 20 and 25 mM) at different water activity (aw) values (0.99, 0.98, 0.96 and 0.93) at 25 °C on growth and fumonisin production by Fusarium verticillioides and Fusarium proliferatum on maize based media. For both Fusarium species, the lag phase significantly decreased (p ≤ 0.001), and the growth rates increased (p ≤ 0.001) at the lowest ferulic acid concentration used (1mM), regardless of the aw. However, high doses of ferulic acid (10 to 25 mM) significantly reduced (p ≤ 0.001) the growth rate of both Fusarium species, regardless of the a(w). In general, growth rate inhibition increased as ferulic acid doses increased and as media aw decreased. Fumonisin production profiles of both Fusarium species showed that low ferulic acid concentrations (1-10mM) significantly increased (p ≤ 0.001) toxin production, regardless of the aw. High doses of ferulic acid (20-25 mM) reduced fumonisin production, in comparison with the controls, by both Fusarium species but they were not statistically significant in most cases. The results show that the use of ferulic acid as a post-harvest strategy to reduce mycotoxin accumulation on maize needs to be discussed.
Product Description
Supelco
4-Hydroxy-3-methoxycinnamic acid, mixture of isomers, analytical standard