MilliporeSigma
  • Pharmacokinetics of 8-O-acetylharpagide and harpagide after oral administration of Ajuga decumbens Thunb extract in rats.
Pharmacokinetics of 8-O-acetylharpagide and harpagide after oral administration of Ajuga decumbens Thunb extract in rats.
Journal of ethnopharmacology (2013-04-03)
Binyu Wen, Rong He, Pengyue Li, Qihua Xu, Yanli Lu, Bo Peng, Jianrong Li
ABSTRACT
Ajuga decumbens Thunb is a medicinal plant native to China popularly used to treat chronic pelvic inflammation and hysteromyoma. Its main bioactive components are iridoid glycosides, such as 8-O-acetylharpagide and harpagide that had presented antibacterial, anti-inflammatory, and antiviral activities. To establish a sensitive LC-MS/MS method and compare the pharmacokinetics of 8-O-acetylharpagide and harpagide in rats after oral administration of their pure forms and from compounds obtained from Ajuga decumbens extract. Rats received orally 15 mg/kg (equivalent of 6 mg/kg 8-O-acetylharpagide and 1.5mg/kg harpagide), 30 mg/kg and 60 mg/kg of Ajuga decumbens Thunb extract and were compared to animals that received 12 mg/kg of 8-O-acetylharpagide or 3mg/kg of harpagide p.o. Concentrations of 8-O-acetylharpagide and harpagide in plasma were determined by LC-MS/MS method at different time points and all pharmacokinetic parameters were estimated by non-compartmental analysis. Results showed that the iridoid glycosides were quickly absorbed by oral route and showed a dose-dependence profile. Pharmacokinetic parameters of both glycosides were essentially the same except Tmax when dosed as the extract or pure forms. 8-O-acetylharpagide was metabolized to harpagide, which affected the pharmacokinetic profiles of harpagide when dosed as the extract. This pharmacokinetic study seems to be useful for a further clinical study of Ajuga decumbens Thunb extract.
Sigma-Aldrich
8-Acetylharpagide, ≥85% (LC/MS-ELSD)