MilliporeSigma
  • The influence of selective vitamin D receptor activator paricalcitol on cardiovascular system and cardiorenal protection.
The influence of selective vitamin D receptor activator paricalcitol on cardiovascular system and cardiorenal protection.
Clinical interventions in aging (2013-02-23)
Darko Duplancic, Marijan Cesarik, Nikola Kolja Poljak, Maja Radman, Vedran Kovacic, Josipa Radic, Veljko Rogosic
ABSTRACT
The ubiquitous distribution of vitamin D receptors in the human body is responsible for the pleiotropic effects of vitamin D-receptor activation. We discuss the possible beneficial effects of a selective activator of vitamin D receptor, paricalcitol, on the cardiovascular system in chronic heart failure patients and chronic kidney patients, in light of new trials. Paricalcitol should provide additional clinical benefits over the standard treatment for chronic kidney and heart failure, especially in cases of cardiorenal syndrome.
Product Description
USP
Paricalcitol, United States Pharmacopeia (USP) Reference Standard