MilliporeSigma
[Biological activity of Penicillium sp. 10-51 exometabolites].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2012-10-24)
Ia I Savchuk, A M Zaĭchenko, E S Tsyganenko
ABSTRACT
Silica gel column chromatography (silica gel "L" II kind of activity 100/160 mkm) of the chloroform extract from the cultural filtrate of Penicillium sp. 10-51 gave two fractions (chloroform and chloroform-acetone, 5:1) having biological activity. Recrystallization yielded two compounds. On the basis of physico-chemical and spectral data these compounds were identified as curvularin and hydroxycurvularin, which have a large spectrum of biological action as to bacteria, yeast, blue-green algae and phytopathogenic micromycetes.
Sigma-Aldrich
Curvularin, from Penicillium citrinum, ≥98% (HPLC)