MilliporeSigma
  • Formulation and pharmacokinetic evaluation of hard gelatin capsule encapsulating lyophilized Vasa Swaras for improved stability and oral bioavailability of vasicine.
Formulation and pharmacokinetic evaluation of hard gelatin capsule encapsulating lyophilized Vasa Swaras for improved stability and oral bioavailability of vasicine.
Fitoterapia (2010-12-29)
Tejas Vyas, Ranjeet Prasad Dash, Sheetal Anandjiwala, Manish Nivsarkar
ABSTRACT
The oral bioavailability of vasicine (1) was investigated in hard gelatin capsules of lyophilized Vasa Swaras (aqueous extract of Adhatoda vasica Nees.,Fam.: Acanthaceae) The rat pharmacokinetic profile of lyophilized Vasa Swaras, Vasa Swaras, vasicine (1) (chief marker compounds of A. vasica) and a marketed capsule formulation of A. vasica were compared. Vasicine (1) was found to be more orally bioavailable from lyophilized Vasa Swaras, with an overall minor conversion to vasicinone (2).