MilliporeSigma
  • Stereoselective analysis of metoprolol and its metabolites in rat plasma with application to oxidative metabolism.
Stereoselective analysis of metoprolol and its metabolites in rat plasma with application to oxidative metabolism.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2005-07-21)
Vanessa Bergamin Boralli, Eduardo Barbosa Coelho, Paula Macedo Cerqueira, Vera Lucia Lanchote
ABSTRACT
We investigated the stereoselective kinetic disposition and metabolism of metoprolol (MET) in rats. The racemic MET (15 mg/kg) was given by oral gavage and blood samples were collected from 0 to 10h (n=6 at each time point). The enantiomeric concentrations of MET and its metabolites alpha-hydroxymetoprolol (alpha-OHM) and O-demethylmetoprolol (ODM) were determined by HPLC using a Chiralpak AD chiral column and fluorescence detection. The pharmacokinetic parameters of unchanged MET and the formation of ODM did not show to be stereoselective. In contrast, the AUC (ng h/mL) of alpha-hydroxymetoprolol isomers were higher to I'R [638.2(525.2-706.2) for 1'R2R and 659.6(580.4-698.1) for 1'R,2S, mean, (95%CI)] than to I'S products [58.3(47.4-66.1) for 1'S,2R and 57.1(44.7-67.9) for 1'S,2S, mean, (95%CI)]. We conclude that the kinetic disposition of unchanged MET and the formation of ODM are not enantioselective in rats but the metabolism of alpha-OHM yields predominantly the 1'R-product.
Product Description
Supelco
α-Hydroxymetoprolol, analytical standard