MilliporeSigma
  • Expeditious asymmetric synthesis of a stereoheptad corresponding to the C(19)-C(27)-ansa chain of rifamycins: formal total synthesis of Rifamycin S.
Expeditious asymmetric synthesis of a stereoheptad corresponding to the C(19)-C(27)-ansa chain of rifamycins: formal total synthesis of Rifamycin S.
Chemistry (Weinheim an der Bergstrasse, Germany) (2004-11-20)
Māris Turks, Xiaogen Huang, Pierre Vogel
ABSTRACT
In the presence of sulfur dioxide and an acid promoter, (-)-(1E,3Z)-2-methyl-1-((1S)-1-phenylethoxy)penta-1,3-dien-3-yl isobutyrate reacts with (Z)-3-(trimethylsilyloxy)pent-2-ene giving a silyl sulfinate intermediate that undergoes, in the presence of palladium catalyst, a desilylation and retro-ene elimination of SO(2) with formation of (-)-(1Z,2S,3R,4S)-1-ethylidene-2,4-dimethyl-5-oxo-3-((1S)-1-phenylethoxy)-heptyl isobutyrate as major product. This ethyl ketone undergoes cross-aldol reaction with (2S)-2-methyl-3-[(tert-butyldimethylsilyl)oxy]propanal giving an aldol that is reduced into a stereoheptad corresponding to the C(19)-C(27)-segment of Rifamycins with high diastereoselectivity and enantiomeric excess.
Product Description
Rifamycin S, European Pharmacopoeia (EP) Reference Standard