MilliporeSigma
  • Pharmacokinetic evaluation of (-)-6-aminocarbovir as a prodrug for (-)-carbovir in rats.
Pharmacokinetic evaluation of (-)-6-aminocarbovir as a prodrug for (-)-carbovir in rats.
Drug metabolism and disposition: the biological fate of chemicals (1992-01-01)
C L Zimmerman, R P Remmel, S S Ibrahim, S A Beers, R Vince
ABSTRACT
The recently synthesized carbocyclic 2',3'-didehydro-2',3'-dideoxy-6-deoxy-6-amino-guanosine [(-)6AC] was evaluated as a prodrug for carbovir, carbocyclic 2',3'-didehydro-2',3'-dideoxyguanosine [(-)CBV] in seven male Sprague-Dawley rats. A randomized three-way cross-over design was used. Rats were assigned to receive the following treatments: a 20 mg/kg (-)6AC infusion, 40 mg/kg (-)6AC orally, and a 20 mg/kg (-)CBV infusion. Blood samples were collected over 480 min, and urine was collected for up to 48 hr. A 2- to 3-day washout period was observed between treatments. Following i.v. infusion, (-)6AC concentrations in the blood declined rapidly in a monoexponential pattern with an elimination half-life of 11.3 +/- 3.3 min (mean +/- SD, n = 7). The time-averaged total body clearance was 115.7 +/- 32.6 ml/min/kg. The fraction of the dose excreted unchanged in urine was 0.28 +/- 0.06. The fraction of the (-)6AC dose metabolized to (-)CBV was 0.48 +/- 0.14. Following oral administration of (-)6AC, the bioavailability of (-)CBV was 46.2 +/- 9.9% (n = 6) in comparison with the bioavailability of approximately 20% previously obtained after an oral dose of (-)CBV. The Cmax of (-)CBV after a 40 mg/kg oral dose of (-)6AC was 1.65 +/- 0.7 micrograms/ml as compared with the previously reported Cmax of 1.00 microgram/ml obtained after a 60 mg/kg oral dose of (-)CBV. (-)6AC has considerable potential for the improvement of the extent of absorption of (-)CBV from oral dosing.
Product Description
USP
Abacavir Related Compound A, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.