MilliporeSigma
Radical deoxygenation of hydroxyl groups via phosphites.
Journal of the American Chemical Society (2004-10-14)
Liming Zhang, Masato Koreeda
ABSTRACT
A highly efficient, two-step sequence method for the deoxygenation of hydroxyl groups has been developed. The method involves the preparation of the 2-(2-iodophenyl)ethyl methyl phosphite derivative of an alcohol using methyl dichlorophosphite and 2-(2-iodophenyl)ethanol. Treatment of the phosphite intermediate with (n-Bu)3SnH/AIBN in refluxing benzene cleanly produces the deoxygenation product of the original alcohol.
Sigma-Aldrich
Methyl dichlorophosphite, technical grade