MilliporeSigma
  • Protective effect of geranylgeranylacetone via enhanced induction of HSPB1 and HSPB8 in mitochondria of the failing heart following myocardial infarction in rats.
Protective effect of geranylgeranylacetone via enhanced induction of HSPB1 and HSPB8 in mitochondria of the failing heart following myocardial infarction in rats.
European journal of pharmacology (2014-03-19)
Tetsuro Marunouchi, Satomi Inomata, Atsushi Sanbe, Norio Takagi, Kouichi Tanonaka
ABSTRACT
The mechanisms underlying mitochondrial impairment in the failing heart are not yet clear. In a previous study, we found that the levels of small heat shock proteins (HSP) such as mitochondrial HSPB1 and HSPB8 in the failing heart following myocardial infarction were decreased. In the present study, to verify the hypothesis that mitochondrial dysfunction in the failing heart is associated with alterations in mitochondrial small heat shock proteins, we examined the effects of geranylgeranylacetone, a heat shock protein inducer, on the cardiac mitochondrial function after myocardial infarction. When hemodynamic parameters of rats with myocardial infarction were measured at the 8th (8W) week after coronary artery ligation (CAL), the 8W-CAL showed signs of chronic heart failure concomitant with a reduced mitochondrial oxygen consumption rate. HSPB1 and HSPB8 contents in the mitochondrial fraction prepared from the failing heart were decreased, suggesting that an attenuation of mitochondrial translocation of HSPB1 and HSPB8 had led to an impairment of mitochondrial energy-producing ability. Geranylgeranylacetone treatment from the 2nd to 8th week after myocardial infarction attenuated the reduction in mitochondrial HSPB1 and HSPB8 contents. Furthermore, the mitochondrial energy-producing ability and cardiac pump function were preserved by orally administered geranylgeranylacetone during the development of heart failure. These results suggest that the induction of small heat shock proteins in the infarcted heart by geranylgeranylacetone treatment contributed to the preservation of mitochondrial function, leading to an improvement of cardiac contractile function.
Sigma-Aldrich
Geranylgeranylacetone
Pricing and availability is not currently available.