MilliporeSigma
  • Vibrational spectroscopic, first-order hyperpolarizability and HOMO, LUMO studies of 4-chloro-2-(trifluoromethyl) aniline based on DFT calculations.
Vibrational spectroscopic, first-order hyperpolarizability and HOMO, LUMO studies of 4-chloro-2-(trifluoromethyl) aniline based on DFT calculations.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-11-15)
M Arivazhagan, V P Subhasini, A Austine
ABSTRACT
The Fourier-transform infrared and FT-Raman spectra of 4-chloro-2-(trifluoromethyl) aniline (4C2TFA) were recorded in the region 4000-400 cm(-1) and 3500-50 cm(-1) respectively. Quantum chemical calculations of energies, geometrical structure and vibrational wavenumbers of 4C2TFA were carried out by density functional theory (DFT/B3LYP) method with 6-311+G(d,p) and 6-311++G(d,p) basis sets. The difference between the observed and scaled wavenumber values of most of the fundamentals is very small. The values of the total dipole moment (μ) and the first order hyperpolarizability (β) of the investigated compound were computed using B3LYP/6-311++G(d,p) calculations. The calculated results also show that 4C2TFA might have microscopic non-linear optical (NLO) behavior with non-zero values. A detailed interpretation of infrared and Raman spectra of 4C2TFA is also reported. The calculated HOMO-LUMO energy gap shows that charge transfer occurs within the molecule.
Sigma-Aldrich
4-Chloro-2-(trifluoromethyl)aniline, 97%
Pricing and availability is not currently available.