MilliporeSigma
Synthesis and properties of Au-Fe3O4 heterostructured nanoparticles.
Journal of colloid and interface science (2012-03-01)
Yang Sheng, Junmin Xue
ABSTRACT
Au-Fe(3)O(4) composite nanoparticles have received much research interest due to their promising biomedical applications. In this work, Au-Fe(3)O(4) composites with well-defined dimer-like nanostructure were synthesized via thermal decomposition route. The surfactant 1,2-hexandicandiol has proved to be critical for the formation of the Au-Fe(3)O(4) hetero-dimers. The hetero-dimers production yield could be significantly improved to be 90% when the 1,2-hexandicandiol concentration was optimized at 0.6 M. The obtained Au-Fe(3)O(4) hetero-dimers possess dual-functionalities of plasmon resonance and magnetization. Moreover, the Fe(3)O(4) domain of the hetero-dimers can be tuned readily by adjusting the molar ratio between Fe and Au sources. Furthermore, it was demonstrated that these Au-Fe(3)O(4) hetero-dimers could be further developed into star-like Au-Fe(3)O(4) nanoparticles which showed plasmon absorption at NIR region.
Sigma-Aldrich
1,2-Hexadecanediol, technical grade, 90%