MilliporeSigma
  • A two-fold increase of carbon cycle sensitivity to tropical temperature variations.
A two-fold increase of carbon cycle sensitivity to tropical temperature variations.
Nature (2014-01-28)
Xuhui Wang, Shilong Piao, Philippe Ciais, Pierre Friedlingstein, Ranga B Myneni, Peter Cox, Martin Heimann, John Miller, Shushi Peng, Tao Wang, Hui Yang, Anping Chen
ABSTRACT
Earth system models project that the tropical land carbon sink will decrease in size in response to an increase in warming and drought during this century, probably causing a positive climate feedback. But available data are too limited at present to test the predicted changes in the tropical carbon balance in response to climate change. Long-term atmospheric carbon dioxide data provide a global record that integrates the interannual variability of the global carbon balance. Multiple lines of evidence demonstrate that most of this variability originates in the terrestrial biosphere. In particular, the year-to-year variations in the atmospheric carbon dioxide growth rate (CGR) are thought to be the result of fluctuations in the carbon fluxes of tropical land areas. Recently, the response of CGR to tropical climate interannual variability was used to put a constraint on the sensitivity of tropical land carbon to climate change. Here we use the long-term CGR record from Mauna Loa and the South Pole to show that the sensitivity of CGR to tropical temperature interannual variability has increased by a factor of 1.9 ± 0.3 in the past five decades. We find that this sensitivity was greater when tropical land regions experienced drier conditions. This suggests that the sensitivity of CGR to interannual temperature variations is regulated by moisture conditions, even though the direct correlation between CGR and tropical precipitation is weak. We also find that present terrestrial carbon cycle models do not capture the observed enhancement in CGR sensitivity in the past five decades. More realistic model predictions of future carbon cycle and climate feedbacks require a better understanding of the processes driving the response of tropical ecosystems to drought and warming.
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Pricing and availability is not currently available.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Pricing and availability is not currently available.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Pricing and availability is not currently available.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Pricing and availability is not currently available.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder
Pricing and availability is not currently available.
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Pricing and availability is not currently available.
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Pricing and availability is not currently available.
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Pricing and availability is not currently available.
Supelco
Activated charcoal, powder
Pricing and availability is not currently available.
Millipore
Activated charcoal, suitable for GC
Pricing and availability is not currently available.
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Pricing and availability is not currently available.
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Pricing and availability is not currently available.
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Pricing and availability is not currently available.
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Pricing and availability is not currently available.
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Pricing and availability is not currently available.
Sigma-Aldrich
Activated charcoal, acid-washed with phosphoric and sulfuric acids
Pricing and availability is not currently available.
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Pricing and availability is not currently available.
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Pricing and availability is not currently available.
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Pricing and availability is not currently available.
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Pricing and availability is not currently available.
Supelco
Activated charcoal, puriss. p.a., powder
Pricing and availability is not currently available.